APO455833027 Apollo Scientific Ltd

3-(2,3-Dihydrobenzo[b]furan-5-yl)propanoic acid

  • MDL number MFCD03102298
  • Cas Number : 215057-28-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144-148
Synonym 5-(2-Carboxyethyl)-2,3-dihydro-1-benzofuran, 3-(2,3-Dihydrobenzo[b]furan-5-yl)propionic acid
Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=27451

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=27451
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart