APO455833043 Apollo Scientific Ltd

2-Phenyl-1,6-naphthyridine-3-carboxylic acid

  • MDL number MFCD04037886
  • Cas Number : 849020-81-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 257-260
Synonym 3-Carboxy-1,6-diaza-2-phenylnaphthalene
Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=27472

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=27472
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart