APO455833045 Apollo Scientific Ltd

4-(4,6-Dimethoxypyrimidin-2-yl)aniline

  • MDL number MFCD01319482
  • Cas Number : 387350-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103-105
Synonym 4-(4,6-Dimethoxy-1,3-diazin-2-yl)aniline, 2-(4-Aminophenyl)-4,6-dimethoxypyrimidine
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-46-dimethoxypyrimidin-2-ylaniline

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-46-dimethoxypyrimidin-2-ylaniline
pictograms GHS06
hstatements H301+H311+H331;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart