APO455833046 Apollo Scientific Ltd

3-[3-(Pyrazin-2-yl)-1,2,4-oxadiazol-5-yl]propanoic acid

  • MDL number MFCD03426086
  • Cas Number : 849925-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >155(dec.)
Synonym 3-[3-(Pyrazin-2-yl)-1,2,4-oxadiazol-5-yl]propionic acid, 5-(2-Carboxyethyl)-3-(pyrazin-2-yl)-1,2,4-oxadiazole
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-3-pyrazin-2-yl-124-oxadiazol-5-ylpropanoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-3-pyrazin-2-yl-124-oxadiazol-5-ylpropanoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart