APO455833050 Apollo Scientific Ltd

4-[3-(Thien-2-yl)-1,2,4-oxadiazol-5-yl]butanoic acid

  • MDL number MFCD03426087
  • Cas Number : 849925-06-0
  • Molecular Weight : 238.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 87-90
Iupac Name 4-[3-(2-thienyl)-1,2,4-oxadiazol-5-yl]butanoic acid
Inchi Code 1S/C10H10N2O3S/c13-9(14)5-1-4-8-11-10(12-15-8)7-3-2-6-16-7/h2-3,6H,1,4-5H2,(H,13,14)
Inchi Key GTNLKFHXTQTTHW-UHFFFAOYSA-N

Description

Synonym 4-[3-(Thien-2-yl)-1,2,4-oxadiazol-5-yl]butyric acid, 5-(3-Carboxyprop-1-yl)-3-(thien-2-yl)-1,2,4-oxadiazole
Purity 95%
Storage temp. Store at 2-8
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 87-90
Product Link https://store.apolloscientific.co.uk/product/4-3-thien-2-yl-124-oxadiazol-5-ylbutanoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-3-thien-2-yl-124-oxadiazol-5-ylbutanoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart