APO455833051 Apollo Scientific Ltd

Ethyl 2,6-dichloro-5-nitropyrimidine-4-carboxylate

  • MDL number MFCD00831638
  • Cas Number : 54368-61-5
  • Molecular Weight : 266.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 36-38
Synonym 2,6-Dichloro-4-(ethoxycarbonyl)-5-nitropyrimidine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-26-dichloro-5-nitropyrimidine-4-carboxylate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-26-dichloro-5-nitropyrimidine-4-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart