APO455833053 Apollo Scientific Ltd

4-(Benzyloxy)-3-methoxyphenylacetonitrile

  • MDL number MFCD00016394
  • Cas Number : 1700-29-4
  • Molecular Weight : 253.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 68-70
Iupac Name [4-(benzyloxy)-3-methoxyphenyl]acetonitrile
Inchi Code 1S/C16H15NO2/c1-18-16-11-13(9-10-17)7-8-15(16)19-12-14-5-3-2-4-6-14/h2-8,11H,9,12H2,1H3
Inchi Key KSOYPRFHKIOHMY-UHFFFAOYSA-N

Description

Synonym 4-(Benzyloxy)-3-methoxybenzyl cyanide, 2-(Benzyloxy)-5-(cyanomethyl)anisole
Purity 98%
Flash Point 195/1mm
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 68-70
Product Link https://store.apolloscientific.co.uk/product/4-benzyloxy-3-methoxyphenylacetonitrile

Safety Information

signalword Danger
pcodes P280;P260
MSDS https://store.apolloscientific.co.uk/product/4-benzyloxy-3-methoxyphenylacetonitrile
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart