APO455833061 Apollo Scientific Ltd

2-Bromo-4-(methylsulphonyl)phenylhydrazine

  • MDL number MFCD04037940
  • Cas Number : 849035-73-0
  • Molecular Weight : 265.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 150-151
Iupac Name 3-bromo-4-hydrazinophenyl methyl sulfone
Inchi Code 1S/C7H9BrN2O2S/c1-13(11,12)5-2-3-7(10-9)6(8)4-5/h2-4,10H,9H2,1H3
Inchi Key CCGWLSRTZZBNTG-UHFFFAOYSA-N

Description

Synonym 2-Bromo-1-hydrazino-4-(methylsulphonyl)benzene, [2-Bromo-4-(methylsulphonyl)phenyl]hydrazine
Purity 95%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 150-151
Product Link https://store.apolloscientific.co.uk/product/2-bromo-4-methylsulphonylphenylhydrazine

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-4-methylsulphonylphenylhydrazine
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart