APO455833067 Apollo Scientific Ltd

1,2-Dimethyl-1H-indole-3-carbonitrile

  • MDL number MFCD00460785
  • Cas Number : 51072-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103-105
Synonym 3-Cyano-1,2-dimethyl-1H-indole
Purity 95
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=27505

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=27505
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart