APO455833068 Apollo Scientific Ltd

(1H-Indol-6-yl)(piperazin-1-yl)methanone

  • MDL number MFCD01765613
  • Cas Number : 633322-11-9
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173-176
Synonym 6-[(Piperazin-1-yl)carbonyl]-1H-indole, 1-[(1H-Indol-6-yl)carbonyl]piperazine
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1h-indol-6-ylpiperazin-1-ylmethanone

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1h-indol-6-ylpiperazin-1-ylmethanone
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart