APO455833069 Apollo Scientific Ltd

1,3-Dihydro-1-(piperidin-4-yl)-2H-indol-2-one

  • MDL number MFCD03426085
  • Cas Number : 16223-25-9
  • Molecular Weight : 216.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 163-166
Iupac Name 1-(4-piperidinyl)-1,3-dihydro-2H-indol-2-one
Inchi Code 1S/C13H16N2O/c16-13-9-10-3-1-2-4-12(10)15(13)11-5-7-14-8-6-11/h1-4,9,11,14,16H,5-8H2
Inchi Key VUEABBGODWKGTK-UHFFFAOYSA-N

Description

Synonym 1-(Piperidin-4-yl)-2-oxindole, 1-(Piperidin-4-yl)indolin-2-one
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 163-166
Product Link https://store.apolloscientific.co.uk/product/13-dihydro-1-piperidin-4-yl-2h-indol-2-one

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/13-dihydro-1-piperidin-4-yl-2h-indol-2-one
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart