APO455833071 Apollo Scientific Ltd

4-(Methylsulphonyl)benzaldehyde

  • MDL number MFCD00216485
  • Cas Number : 5398-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-161
Synonym 4-Formylphenyl methyl sulphone
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-methylsulphonylbenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-methylsulphonylbenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart