APO455833074 Apollo Scientific Ltd

1-(3-Nitropyridin-2-yl)piperidine-4-carbonitrile

  • MDL number MFCD00106383
  • Cas Number : 671191-99-4
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 98-100
Iupac Name 1-(3-nitro-2-pyridinyl)-4-piperidinecarbonitrile
Inchi Code 1S/C11H12N4O2/c12-8-9-3-6-14(7-4-9)11-10(15(16)17)2-1-5-13-11/h1-2,5,9H,3-4,6-7H2
Inchi Key UEXODVNTPRVAEL-UHFFFAOYSA-N

Description

Synonym 2-(4-Cyanopiperidin-1-yl)-3-nitropyridine, 4-Cyano-1-(3-nitropyridin-2-yl)piperidine
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 98-100
Product Link https://store.apolloscientific.co.uk/product/1-3-nitropyridin-2-ylpiperidine-4-carbonitrile

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-3-nitropyridin-2-ylpiperidine-4-carbonitrile
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart