APO455833078 Apollo Scientific Ltd

1-Phenyl-2-(pyrimidin-4-yl)ethan-1-one

  • MDL number MFCD00559708
  • Cas Number : 36912-83-1
  • Molecular Weight : 198.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-phenyl-2-(4-pyrimidinyl)ethanone
Inchi Code 1S/C12H10N2O/c15-12(10-4-2-1-3-5-10)8-11-6-7-13-9-14-11/h1-7,9H,8H2
Inchi Key BJDUPABHTHYHEX-UHFFFAOYSA-N

Description

Synonym 4-(Benzoylmethyl)-1,3-diazine, 4-Phenacylpyrimidine
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-phenyl-2-pyrimidin-4-ylethan-1-one

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-phenyl-2-pyrimidin-4-ylethan-1-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart