APO455833079 Apollo Scientific Ltd

4-Bromo-2-(methylsulphonyl)benzyl bromide

  • MDL number MFCD04037945
  • Cas Number : 254887-18-8
  • Molecular Weight : 328.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 124-128
Iupac Name 4-bromo-1-(bromomethyl)-2-(methylsulfonyl)benzene
Inchi Code 1S/C8H8Br2O2S/c1-13(11,12)8-4-7(10)3-2-6(8)5-9/h2-4H,5H2,1H3
Inchi Key PJFOHHNHJUOESV-UHFFFAOYSA-N

Description

Synonym 4-Bromo-1-(bromomethyl)-2-(methylsulphonyl)benzene, 5-Bromo-2-(bromomethyl)phenyl methyl sulphone, alpha,4-Dibromo-2-(methylsulphonyl)toluene
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 124-128
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-methylsulphonylbenzyl-bromide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-methylsulphonylbenzyl-bromide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart