APO455833082 Apollo Scientific Ltd

4-Bromo-2-(methylsulphonyl)toluene

  • MDL number MFCD04037925
  • Cas Number : 254887-17-7
  • Molecular Weight : 249.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 92-94
Iupac Name 4-bromo-1-methyl-2-(methylsulfonyl)benzene
Inchi Code 1S/C8H9BrO2S/c1-6-3-4-7(9)5-8(6)12(2,10)11/h3-5H,1-2H3
Inchi Key SRYXZVWRMOFVCO-UHFFFAOYSA-N

Description

Synonym 5-Bromo-2-methylphenyl methyl sulphone, 4-Bromo-1-methyl-2-(methylsulphonyl)benzene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 92-94
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-methylsulphonyltoluene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-methylsulphonyltoluene
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart