APO455833096 Apollo Scientific Ltd

Pyrimidine-4-carboxaldehyde

  • MDL number MFCD03789624
  • Cas Number : 2435-50-9
  • Molecular Weight : 108.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-pyrimidinecarbaldehyde
Inchi Code 1S/C5H4N2O/c8-3-5-1-2-6-4-7-5/h1-4H
Inchi Key OKULHRWWYCFJAB-UHFFFAOYSA-N

Description

Synonym 4-Formylpyrimidine, 4-Formyl-1,3-diazine
Purity 95%
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/pyrimidine-4-carboxaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/pyrimidine-4-carboxaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart