APO455833097 Apollo Scientific Ltd

N-Methyl-4-(methylsulphonyl)-2-nitroaniline

  • MDL number MFCD01016949
  • Cas Number : 30388-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188-190
Synonym Methyl 4-(methylamino)-3-nitrophenyl sulphone, 2-(Methylamino)-5-(methylsulphonyl)nitrobenzene
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=27560

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=27560
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart