APO455833098 Apollo Scientific Ltd

3-(Methylsulphonyl)benzenesulphonyl chloride

  • MDL number MFCD04037968
  • Cas Number : 5335-40-0
  • Molecular Weight : 254.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 93-96
Iupac Name 3-(methylsulfonyl)benzenesulfonyl chloride
Inchi Code 1S/C7H7ClO4S2/c1-13(9,10)6-3-2-4-7(5-6)14(8,11)12/h2-5H,1H3
Inchi Key CQEFJGDLGWJIRV-UHFFFAOYSA-N

Description

Synonym 3-(Chlorosulphonyl)phenyl methyl sulphone
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 93-96
Product Link https://store.apolloscientific.co.uk/product/3-methylsulphonylbenzenesulphonyl-chloride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-methylsulphonylbenzenesulphonyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart