APO455833104 Apollo Scientific Ltd

5-Amino-1H-indole-3-carboxamide hydrochloride

  • MDL number MFCD02259416
  • Cas Number : 306936-36-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 296-298
Synonym 3-(Aminocarbonyl)-1H-indol-5-amine hydrochloride
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=12670

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=12670
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart