APO455833109 Apollo Scientific Ltd

4-[(1H-Benzimidazol-1-yl)methyl]benzoic acid

  • MDL number MFCD02180372
  • Cas Number : 139742-50-0
  • Molecular Weight : 252.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 260-265
Synonym 1-(4-Carboxybenzyl)-1H-benzimidazole, 1-[(4-Carboxyphenyl)methyl]-1H-benzimidazole
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-1h-benzimidazol-1-ylmethylbenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-1h-benzimidazol-1-ylmethylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart