APO455833110 Apollo Scientific Ltd

(1,2-Benzisoxazol-3-yl)acetic acid

  • MDL number MFCD02180388
  • Cas Number : 4865-84-3
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123-129
Synonym 3-(Carboxymethyl)-1,2-benzisoxazole, (Benzo[d]isoxazol-3-yl)acetic acid
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/12-benzisoxazol-3-ylacetic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/12-benzisoxazol-3-ylacetic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart