APO455833120 Apollo Scientific Ltd

5-(Bromoacetyl)-1,3-benzodioxole

  • MDL number MFCD01910077
  • Cas Number : 40288-65-1
  • Molecular Weight : 243.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91-94
Synonym 1-(1,3-Benzodioxol-5-yl)-2-bromoethan-1-one, 3,4-(Methylenedioxy)phenacyl bromide
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromoacetyl-13-benzodioxole

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromoacetyl-13-benzodioxole
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart