APO455833133 Apollo Scientific Ltd

5-Chloro-1-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD02089694
  • Cas Number : 54367-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197-198
Synonym 4-Carboxy-5-chloro-1-methyl-1H-pyrazole
Purity 94
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13819

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13819
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart