APO455833136 Apollo Scientific Ltd

2-(5-Chloro-1,2,4-thiadiazol-3-yl)thiophene-5-sulphonyl chloride

  • MDL number MFCD02180888
  • Cas Number : 306937-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71-74
Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13940

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13940
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart