APO455833144 Apollo Scientific Ltd

Ethyl [(4-iodophenyl)amino]acetate

  • MDL number MFCD00176764
  • Cas Number : 14108-76-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84-86
Synonym Ethyl 2-(4-iodoanilino)acetate
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=14658

Safety Information

signalword Warning
pcodes P280;P261
MSDS http://www.apolloscientific.co.uk/display_item.php?id=14658
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart