APO455833146 Apollo Scientific Ltd

4-Amino-1,3,5-trimethyl-1H-pyrazole

  • MDL number MFCD00052883
  • Cas Number : 28466-21-9
  • Molecular Weight : 125.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 99-105
Iupac Name 1,3,5-trimethyl-1H-pyrazol-4-amine
Inchi Code 1S/C6H11N3/c1-4-6(7)5(2)9(3)8-4/h7H2,1-3H3
Inchi Key SSDGMKHZMNTWLS-UHFFFAOYSA-N

Description

Synonym 1,3,5-Trimethyl-1H-pyrazol-4-amine
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 99-105
Product Link https://store.apolloscientific.co.uk/product/4-amino-135-trimethyl-1h-pyrazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-135-trimethyl-1h-pyrazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart