APO455833150 Apollo Scientific Ltd

4-(4-Methoxyphenyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine

  • MDL number MFCD00177096
  • Cas Number : 213462-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69-71
Synonym 4-(4,5,6,7-Tetrahydrothieno[3,2-c]pyridin-4-yl)anisole
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=15096

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=15096
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart