APO455833157 Apollo Scientific Ltd

5-(N,N-Dimethylsulphamoyl)-2-methyl-3-furoic acid

  • MDL number MFCD02180761
  • Cas Number : 306936-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169
Synonym 5-(N,N-Dimethylsulphamoyl)-2-methylfuran-3-carboxylic acid, 5-[(Dimethylamino)sulphonyl]-2-methylfuran-3-carboxylic acid
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=15197

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=15197
pictograms GHS05
hstatements H314;H318
Availability details for
Quantity
  • Availability
  • Add to Cart