APO455833159 Apollo Scientific Ltd

1-Methyl-1H-imidazole-4-sulphonamide

  • MDL number MFCD02091382
  • Cas Number : 111124-90-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 222-224
Synonym 1-Methyl-4-sulphamoyl-1H-imidazole, 4-(Aminosulphonyl)-1-methyl-1H-imidazole
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=15222

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=15222
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart