APO455833162 Apollo Scientific Ltd

2-Methyl-5-(piperidin-1-ylsulphonyl)-3-furoic acid

  • MDL number MFCD02180759
  • Cas Number : 306936-38-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188-189
Synonym 2-Methyl-5-(piperidin-1-ylsulphonyl)furan-3-carboxylic acid
Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=15289

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=15289
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart