APO455833164 Apollo Scientific Ltd

2-(Morpholin-4-yl)benzaldehyde

  • MDL number MFCD00662562
  • Cas Number : 58028-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45-48
Synonym 4-(2-Formylphenyl)morpholine
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 162/12mm
Product Link https://store.apolloscientific.co.uk/product/2-morpholin-4-ylbenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-morpholin-4-ylbenzaldehyde
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart