APO455833172 Apollo Scientific Ltd

Methyl 4-bromo-3-nitrobenzoate

  • MDL number MFCD00553089
  • Cas Number : 2363-16-8
  • Molecular Weight : 260.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 104
Iupac Name methyl 4-bromo-3-nitrobenzoate
Inchi Code 1S/C8H6BrNO4/c1-14-8(11)5-2-3-6(9)7(4-5)10(12)13/h2-4H,1H3
Inchi Key BNNDHGPPQZVKMX-UHFFFAOYSA-N

Description

Synonym 2-Bromo-5-(methoxycarbonyl)nitrobenzene
Storage temp. Ambient Temperature
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 104
Product Link https://store.apolloscientific.co.uk/product/methyl-4-bromo-3-nitrobenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-4-bromo-3-nitrobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart