APO455833174 Apollo Scientific Ltd

5-(4-Bromobut-1-yl)hydantoin

  • MDL number MFCD00832189
  • Cas Number : 28484-49-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129-131
Synonym 5-(4-Bromobut-1-yl)imidazolidine-2,4-dione, 5-(4-Bromobut-1-yl)-2,4-dioxoimidazolidine
Storage temp. Ambient temperature
Physical Form Solid-Crystals
Shipping Temperature Shipped with Ice Packs
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13180

Safety Information

signalword Warning
pcodes P271;P280;P261
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13180
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart