APO455833176 Apollo Scientific Ltd

4'-(Bromomethyl)-3'-chloro-[1,1'-biphenyl]-2-carbonitrile

  • MDL number MFCD07777122
  • Cas Number : 887268-24-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chloro-4-(2-cyanophenyl)benzyl bromide, 2-(4-(Bromomethyl)-3-chlorophenyl)benzonitrile
Purity 98
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=32524

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=32524
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart