APO455833178 Apollo Scientific Ltd

1-Benzyl-4-methyl-1H-1,2,3-triazole

  • MDL number MFCD08062394
  • Cas Number : 91258-00-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym [(4-Methyl-1H-1,2,3-triazol-1-yl)methyl]benzene, alpha-(4-Methyl-1H-1,2,3-triazol-1-yl)toluene
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36059

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=36059
pictograms GHS02;GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart