APO455833187 Apollo Scientific Ltd

4-(4-Methylphenyl)pyrimidin-2-amine

  • MDL number MFCD00665911
  • Cas Number : 263276-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Amino-4-(4-methylphenyl)pyrimidine, 4-(2-Aminopyrimidin-4-yl)toluene, 2-Amino-4-(p-tolyl)-1,3-diazine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=12720

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=12720
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart