APO455833192 Apollo Scientific Ltd

3-Hydroxy-5-nitrobenzonitrile

  • MDL number MFCD09258662
  • Cas Number : 929000-02-2
  • Molecular Weight : 164.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Cyano-5-nitrophenol, 3-Cyano-5-hydroxynitrobenzene
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-hydroxy-5-nitrobenzonitrile

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-hydroxy-5-nitrobenzonitrile
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart