APO455833208 Apollo Scientific Ltd

8-Methylquinazolin-4(3H)-one

  • MDL number MFCD01685945
  • Cas Number : 19181-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 8-Methylquinazolin-4-ol
Purity 98
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/8-methylquinazolin-43h-one

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/8-methylquinazolin-43h-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart