APO455833209 Apollo Scientific Ltd

5,8-Dibromoisoquinoline

  • MDL number MFCD03109888
  • Cas Number : 81045-39-8
  • Molecular Weight : 286.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5,8-dibromoisoquinoline
Inchi Code 1S/C9H5Br2N/c10-8-1-2-9(11)7-5-12-4-3-6(7)8/h1-5H
Inchi Key KNBALCRXZUTMBP-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/58-dibromoisoquinoline

Safety Information

signalword Warning
pcodes P280
MSDS https://store.apolloscientific.co.uk/product/58-dibromoisoquinoline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart