APO455833210 Apollo Scientific Ltd

2-Amino-3-bromo-5-chloropyridine

  • MDL number MFCD00955627
  • Cas Number : 26163-03-1
  • Molecular Weight : 207.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 83-85
Iupac Name 3-bromo-5-chloro-2-pyridinylamine
Inchi Code 1S/C5H4BrClN2/c6-4-1-3(7)2-9-5(4)8/h1-2H,(H2,8,9)
Inchi Key UWGGGYYCKDCTGN-UHFFFAOYSA-N

Description

Synonym 3-Bromo-5-chloropyridin-2-amine
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 83-85
Product Link https://store.apolloscientific.co.uk/product/2-amino-3-bromo-5-chloropyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-3-bromo-5-chloropyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart