APO455833211 Apollo Scientific Ltd

6-Chloro-1,3-benzothiazole-2(3H)-thione

  • MDL number MFCD02677573
  • Cas Number : 51618-29-2
  • Molecular Weight : 201.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-chloro-1,3-benzothiazole-2(3H)-thione
Inchi Code 1S/C7H4ClNS2/c8-4-1-2-5-6(3-4)11-7(10)9-5/h1-3H,(H,9,10)
Inchi Key CLHLOHAQAADLRA-UHFFFAOYSA-N

Description

Synonym 6-Chloro-2,3-dihydro-2-thioxo-1,3-benzothiazole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-chloro-13-benzothiazole-23h-thione

Safety Information

signalword Warning
pcodes P271;P280;P261
MSDS https://store.apolloscientific.co.uk/product/6-chloro-13-benzothiazole-23h-thione
pictograms GHS07
hstatements H302;H312;H315;H319;H332;H335
Availability details for
Quantity
  • Availability
  • Add to Cart