APO455833225 Apollo Scientific Ltd

2-Chloro-4-(hydroxymethyl)-1,3-thiazole

  • MDL number MFCD09265529
  • Cas Number : 5198-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (2-Chloro-1,3-thiazol-4-yl)methanol
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 46/10mm
Product Link https://store.apolloscientific.co.uk/product/2-chloro-4-hydroxymethyl-13-thiazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-4-hydroxymethyl-13-thiazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart