APO455833228 Apollo Scientific Ltd

4,5-Dimethyl-2-(tributylstannyl)-1,3-thiazole

  • MDL number MFCD09265485
  • Cas Number : 938181-92-1
  • Molecular Weight : 402.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4,5-dimethyl-2-(tributylstannyl)-1,3-thiazole
Inchi Code 1S/C5H6NS.3C4H9.Sn/c1-4-5(2)7-3-6-4;3*1-3-4-2;/h1-2H3;3*1,3-4H2,2H3;
Inchi Key QAPTXJNMSZQTDL-UHFFFAOYSA-N

Description

Synonym (4,5-Dimethyl-1,3-thiazol-2-yl)tributylstannane
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/45-dimethyl-2-tributylstannyl-13-thiazole

Safety Information

signalword Danger
pcodes P280
MSDS https://store.apolloscientific.co.uk/product/45-dimethyl-2-tributylstannyl-13-thiazole
pictograms GHS06
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart