APO455833230 Apollo Scientific Ltd

4-Aminotetrahydro-2H-thiopyran

  • MDL number MFCD07371524
  • Cas Number : 21926-00-1
  • Molecular Weight : 117.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tetrahydro-2H-thiopyran-4-amine
Inchi Code 1S/C5H11NS/c6-5-1-3-7-4-2-5/h5H,1-4,6H2
Inchi Key RPIXOLUIHUFDOY-UHFFFAOYSA-N

Description

Synonym Tetrahydro-2H-thiopyran-4-amine, 4-Aminothiane
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-aminotetrahydro-2h-thiopyran

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-aminotetrahydro-2h-thiopyran
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart