APO455833231 Apollo Scientific Ltd

1H-Pyrazole-5-boronic acid

  • MDL number MFCD07368247
  • Cas Number : 376584-63-3
  • Molecular Weight : 111.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1H-pyrazol-5-ylboronic acid
Inchi Code 1S/C3H5BN2O2/c7-4(8)3-1-2-5-6-3/h1-2,7-8H,(H,5,6)
Inchi Key NEUWPDLMDVINSN-UHFFFAOYSA-N

Description

Synonym 2H-Pyrazole-3-boronic acid, 5-Borono-1H-pyrazole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1h-pyrazole-5-boronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1h-pyrazole-5-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart