APO455833233 Apollo Scientific Ltd

Propylboronic acid

  • MDL number MFCD01074564
  • Cas Number : 17745-45-8
  • Molecular Weight : 87.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 105-108
Iupac Name propylboronic acid
Inchi Code 1S/C3H9BO2/c1-2-3-4(5)6/h5-6H,2-3H2,1H3
Inchi Key JAQOMSTTXPGKTN-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 105-108
Product Link https://store.apolloscientific.co.uk/product/propylboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/propylboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart