APO455833238 Apollo Scientific Ltd

3-Bromo-2-methoxy-5-nitropyridine

  • MDL number MFCD06659517
  • Cas Number : 15862-50-7
  • Molecular Weight : 233.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 89
Iupac Name 3-bromo-2-methoxy-5-nitropyridine
Inchi Code 1S/C6H5BrN2O3/c1-12-6-5(7)2-4(3-8-6)9(10)11/h2-3H,1H3
Inchi Key VRCSBPXFYAADHQ-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 89
Product Link https://store.apolloscientific.co.uk/product/3-bromo-2-methoxy-5-nitropyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-2-methoxy-5-nitropyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart