APO455833249 Apollo Scientific Ltd

Ethyl 5-aminobenzo[b]furan-2-carboxylate

  • MDL number MFCD08275091
  • Cas Number : 174775-48-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54-65
Synonym 5-Amino-2-(ethoxycarbonyl)-1-benzofuran, Ethyl 5-amino-1-benzofuran-2-carboxylate
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36634

Safety Information

signalword Warning
pcodes P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=36634
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart