APO455833254 Apollo Scientific Ltd

4-(3-Bromophenyl)morpholine

  • MDL number MFCD04112585
  • Cas Number : 197846-82-5
  • Molecular Weight : 242.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(3-bromophenyl)morpholine
Inchi Code 1S/C10H12BrNO/c11-9-2-1-3-10(8-9)12-4-6-13-7-5-12/h1-3,8H,4-7H2
Inchi Key RQMIVKDXRGZOBQ-UHFFFAOYSA-N

Description

Synonym 1-Bromo-3-(morpholin-4-yl)benzene
Purity 96%
Storage temp. Ambient Temperature
Physical Form Liquid-Viscous
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-3-bromophenylmorpholine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-3-bromophenylmorpholine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart